siRNA Details
Gene Symbol: TSPYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022117.3 | NP_071400.1 | 1 | 642 | ||
XM_006724592.3 | XP_006724655.1 | 1 | 675 | |||
XM_017029726.1 | XP_016885215.1 | 1 | 675 | |||
XM_017029727.1 | XP_016885216.1 | 1 | 675 | |||
GenBank mRNA | AF254794.1 | - | 1 | 381 | ||
AF273046.1 | - | 1 | 632 | |||
AK131220.1 | - | 1 | 635 | |||
AK292462.1 | - | 1 | 635 | |||
AL713652.1 | - | 1 | 621 | |||
AY040871.1 | - | 1 | 631 | |||
BC024270.1 | - | 1 | 622 | 2819684 |
