siRNA Details
Gene Symbol: NKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024528.3 | NP_078804.2 | 7, 8 | 1091 | ||
XM_017029842.1 | XP_016885331.1 | 7, 8 | 628 | |||
GenBank mRNA | AK026279.1 | - | 7, 8 | 1091 | ||
AK094446.1 | - | 5, 6 | 1852 | |||
AL832118.1 | - | 9, 10 | 1032 | |||
AY388958.1 | - | 7, 8 | 1091 | |||
BC000940.2 | - | 7, 8 | 973 | |||
BC015354.1 | - | 7, 8 | 955 | 3450847 | ||
BC032442.1 | - | 7, 8 | 949 | 5139142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
