siRNA Details
Gene Symbol: EFCAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024593.3 | NP_078869.1 | 6 | 846 | ||
XM_011517590.2 | XP_011515892.1 | 7 | 976 | |||
XM_011517591.2 | XP_011515893.1 | 7 | 949 | |||
XM_017013827.1 | XP_016869316.1 | 8 | 1099 | |||
GenBank mRNA | AK021829.1 | - | 6 | 704 | ||
AK316037.1 | - | 5 | 642 | |||
BC025676.1 | - | 6 | 767 | 5203436 |
