siRNA Details
Gene Symbol: ZMAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135731.1 | NP_001129203.1 | 3 | 273 | ||
NM_024645.2 | NP_078921.1 | 3 | 273 | |||
XM_017013836.1 | XP_016869325.1 | 3 | 277 | |||
XM_017013837.1 | XP_016869326.1 | 4 | 331 | |||
XM_017013838.1 | XP_016869327.1 | 4 | 369 | |||
XM_017013839.1 | XP_016869328.1 | 4 | 272 | |||
GenBank mRNA | AK023904.1 | - | 3 | 273 | ||
BC019598.1 | - | 3 | 259 | 4934088 |
