siRNA Details
Gene Symbol: ALG13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257230.1 | NP_001244159.1 | 23, 24 | 2695 | ||
NM_001257234.1 | NP_001244163.1 | 23, 24 | 2768 | |||
NM_001257237.1 | NP_001244166.1 | 23, 24 | 2872 | |||
NM_001324292.1 | NP_001311221.1 | 23, 24 | 2799 | |||
NM_001324293.1 | NP_001311222.1 | 21, 22 | 2698 | |||
XM_011531030.1 | XP_011529332.1 | 24, 25 | 3243 | |||
XM_017029848.1 | XP_016885337.1 | 23, 24 | 3147 | |||
XM_017029855.1 | XP_016885344.1 | 16, 17 | 2437 | |||
GenBank mRNA | AK026671.1 | - | 8, 9 | 928 | ||
AK223154.1 | - | 8, 9 | 928 | |||
AK300394.1 | - | 23, 24 | 2637 | |||
AK316306.1 | - | 23, 24 | 2741 | |||
AK316522.1 | - | 23, 24 | 2816 | |||
BC117377.1 | - | 8, 9 | 926 | 40125928 | ||
BC117379.1 | - | 8, 9 | 926 | 40125930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
