siRNA Details
Gene Symbol: ADGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025048.3 | NP_079324.2 | 8 | 1301 | ||
GenBank mRNA | AK026337.1 | - | 8 | 1232 | ||
AK225460.1 | - | 8 | 1232 | |||
CR627234.1 | - | 7 | 1138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
