siRNA Details
Gene Symbol: ABHD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319305.1 | NP_001306234.1 | 11 | 1420 | ||
NM_001319306.1 | NP_001306235.1 | 10 | 1375 | |||
NM_001319307.1 | NP_001306236.1 | 11 | 1438 | |||
XM_005263238.4 | XP_005263295.1 | 11 | 1471 | |||
XM_006714318.3 | XP_006714381.1 | 12 | 1516 | |||
XM_006714320.3 | XP_006714383.1 | 12 | 1304 | |||
XM_006714322.3 | XP_006714385.1 | 8 | 854 | |||
XM_011532272.2 | XP_011530574.1 | 13 | 1583 | |||
XM_011532273.2 | XP_011530575.1 | 12 | 1538 | |||
XM_011532274.2 | XP_011530576.1 | 11 | 1414 | |||
XM_011532276.1 | XP_011530578.1 | 11 | 1414 | |||
XM_011532278.2 | XP_011530580.1 | 10 | 1277 | |||
XM_011532279.2 | XP_011530581.1 | 5 | 790 | |||
XM_017008631.1 | XP_016864120.1 | 10 | 1369 | |||
XM_017008632.1 | XP_016864121.1 | 5 | 704 | |||
GenBank mRNA | AK024759.1 | - | 10 | 1187 | ||
BC034253.1 | - | 10 | 1253 | 4339231 | ||
BC111400.1 | - | 10 | 1355 | |||
BC128142.1 | - | 10 | 1125 | 40112521 | ||
BC128143.1 | - | 10 | 1068 | 40112522 |
