siRNA Details
Gene Symbol: RNF170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160223.1 | NP_001153695.1 | 3 | 659 | ||
NM_001160224.1 | NP_001153696.1 | 3 | 429 | |||
NM_001160225.1 | NP_001153697.1 | 3 | 308 | |||
NM_030954.3 | NP_112216.3 | 3 | 429 | |||
NR_027668.1 | 3 | 659 | ||||
NR_027669.1 | 3 | 429 | ||||
XM_006716404.2 | XP_006716467.1 | 4 | 416 | |||
XM_011544666.2 | XP_011542968.1 | 3 | 213 | |||
XM_017013881.1 | XP_016869370.1 | 4 | 356 | |||
GenBank mRNA | AK027748.1 | - | 3 | 345 | ||
AK090864.1 | - | 3 | 592 | |||
AK225086.1 | - | 3 | 309 | |||
AK225568.1 | - | 3 | 325 | |||
AL136620.1 | - | 3 | 313 | |||
BC013422.1 | - | 3 | 294 | |||
BC032393.1 | - | 3 | 291 | 5200847 | ||
BC039461.1 | - | 3 | 416 | |||
BC044566.1 | - | 3 | 429 | |||
BC058289.1 | - | 3 | 292 | |||
BG910500.1 | - | 3 | 650 | |||
DA751961.1 | - | 3 | 429 |
