siRNA Details
Gene Symbol: TRAPPC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031466.7 | NP_113654.4 | 1 | 272 | ||
XM_011517326.2 | XP_011515628.1 | 1 | 272 | |||
XM_011517328.2 | XP_011515630.1 | 1 | 272 | |||
XM_017013893.1 | XP_016869382.1 | 1 | 272 | |||
GenBank mRNA | AK128755.1 | - | 1 | 273 |
