siRNA Details
Gene Symbol: TPD52L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033516.5 | NP_277051.3 | 1 | 917 | ||
GenBank mRNA | AK057479.1 | - | 1 | 891 | ||
AY032877.1 | - | 1 | 895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
