siRNA Details
Gene Symbol: FAM92A1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003612.2 | 1 | 386 | |||
GenBank mRNA | BC022019.1 | - | 1 | 349 | ||
BC047622.1 | - | 1 | 386 | |||
BC059396.1 | - | 1 | 386 | |||
BC094692.1 | - | 1 | 351 |
Gene Symbol: FAM92A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283034.1 | NP_001269963.1 | 3 | 426 | ||
NM_145269.4 | NP_660312.2 | 3 | 426 | |||
NR_104267.1 | 3 | 426 | ||||
NR_104268.1 | 3 | 426 | ||||
XM_005250787.2 | XP_005250844.1 | 3 | 296 | |||
GenBank mRNA | AK096298.1 | - | 3 | 320 | ||
BC014598.2 | - | 3 | 426 | |||
BC108707.1 | - | 3 | 388 | |||
CR627475.1 | - | 3 | 1465 | |||
DQ327715.1 | - | 3 | 340 | |||
DQ327716.1 | - | 3 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
