siRNA Details
Gene Symbol: RNF182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165032.1 | NP_001158504.1 | 4 | 2873 | ||
NM_001165033.1 | NP_001158505.1 | 2 | 2652 | |||
NM_001165034.1 | NP_001158506.1 | 3 | 2760 | |||
NM_152737.3 | NP_689950.1 | 3 | 2790 | |||
XM_017010448.1 | XP_016865937.1 | 4 | 2810 | |||
XM_017010449.1 | XP_016865938.1 | 3 | 2874 | |||
XM_017010450.1 | XP_016865939.1 | 2 | 2858 | |||
GenBank mRNA | AK090576.1 | - | 2 | 2609 | ||
AK098091.1 | - | 1 | 1464 | |||
BC030666.1 | - | 3 | 2690 | 5259219 | ||
BC050030.1 | - | 2 | 2511 | 5297801 |
