siRNA Details
Gene Symbol: HIPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113239.2 | NP_001106710.1 | 2 | 723 | ||
NM_022740.4 | NP_073577.3 | 2 | 723 | |||
XM_006715935.2 | XP_006715998.1 | 3 | 712 | |||
XM_011516077.2 | XP_011514379.1 | 2 | 1301 | |||
XM_011516078.2 | XP_011514380.1 | 2 | 1302 | |||
XM_011516079.2 | XP_011514381.1 | 2 | 1301 | |||
XM_011516080.2 | XP_011514382.1 | 2 | 1302 | |||
XM_011516081.2 | XP_011514383.1 | 2 | 698 | |||
XM_017012069.1 | XP_016867558.1 | 2 | 804 | |||
GenBank mRNA | AF207702.1 | - | 1 | 431 | ||
AF208291.1 | - | 2 | 560 | |||
AF326592.1 | - | 2 | 607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
