siRNA Details
Gene Symbol: TMEM196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005249692.4 | XP_005249749.2 | 4 | 1497 | ||
XM_011515249.2 | XP_011513551.1 | 4 | 1479 | |||
XM_017011928.1 | XP_016867417.1 | 4 | 1497 | |||
XM_017011929.1 | XP_016867418.1 | 4 | 1479 | |||
GenBank mRNA | BC030104.1 | - | 4 | 358 | 4797383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
