siRNA Details
Gene Symbol: CERK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022766.5 | NP_073603.2 | 3 | 418 | ||
XM_017028909.1 | XP_016884398.1 | 3 | 215 | |||
GenBank mRNA | AB051433.1 | - | 2 | 144 | ||
AB079066.1 | - | 3 | 434 | |||
AJ457828.1 | - | 3 | 381 | |||
AK291476.1 | - | 3 | 374 | |||
BC067255.1 | - | 3 | 370 | |||
BC126940.1 | - | 3 | 361 | 40084110 | ||
CR456404.1 | - | 3 | 321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
