siRNA Details
Gene Symbol: XKR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173683.3 | NP_775954.2 | 3 | 2950 | ||
GenBank mRNA | BC024146.2 | - | 3 | 2253 | 3678468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
