siRNA Details
Gene Symbol: NDUFAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174889.4 | NP_777549.1 | 4 | 632 | ||
GenBank mRNA | AF087990.1 | - | 4 | 601 | ||
AK291296.1 | - | 4 | 595 | |||
BC001753.1 | - | 1 | 554 | 3354845 | ||
BC033965.1 | - | 4 | 548 | 5288040 | ||
BC070357.1 | - | 3 | 510 |
