siRNA Details
Gene Symbol: NADK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198994.1 | NP_001185923.1 | 7 | 1009 | ||
XM_017002145.1 | XP_016857634.1 | 5 | 835 | |||
XM_017002146.1 | XP_016857635.1 | 5 | 705 | |||
GenBank mRNA | AF250320.1 | - | 7 | 1009 | ||
AK307645.1 | - | 4 | 647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
