siRNA Details
Gene Symbol: CTAGE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008747.2 | NP_001008747.1 | 1 | 250 | ||
GenBank mRNA | BC146818.1 | - | 1 | 250 | 8860423 |
Gene Symbol: CTAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198495.2 | NP_940897.2 | 1 | 282 | ||
GenBank mRNA | DB727739.1 | - | 1 | 282 | ||
EL733901.1 | - | 1 | 98 |
Gene Symbol: CTAGE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178561.4 | NP_848656.2 | 1 | 325 |
Gene Symbol: CTAGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278507.1 | NP_001265436.1 | 1 | 767 | ||
GenBank mRNA | AK292236.1 | - | 1 | 271 |
Gene Symbol: CTAGE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145659.1 | NP_001139131.1 | 1 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
