siRNA Details
Gene Symbol: RSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178565.4 | NP_848660.3 | 2, 3 | 756 | ||
XM_017013395.1 | XP_016868884.1 | 2, 3 | 745 | |||
GenBank mRNA | AK123023.1 | - | 2, 3 | 719 | ||
AY358166.1 | - | 2, 3 | 409 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172815, BC052844, AK087485, AK052457, AK049891.
Gene Ontology Categories:
