siRNA Details
Gene Symbol: AKAP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178813.5 | NP_848928.1 | 4 | 457 | ||
GenBank mRNA | AF514780.1 | - | 4 | 404 | ||
BC066357.1 | - | 5 | 594 | 5295545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
