siRNA Details
Gene Symbol: LRRC70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181506.4 | NP_852607.3 | 2 | 1959 | ||
GenBank mRNA | AK123022.1 | - | 2 | 1959 | ||
AY280614.1 | - | 2 | 1804 | |||
AY358733.1 | - | 2 | 1823 | |||
BC067275.1 | - | 1 | 1488 | |||
BC110838.1 | - | 2 | 1811 |

More Information
Other Species Targeted

Gene Ontology Categories:
