siRNA Details
Gene Symbol: RPL34-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026968.1 | 2 | 309 | |||
GenBank mRNA | AK094992.1 | - | 2 | 249 | ||
AK298931.1 | - | 2 | 281 | |||
AK308624.1 | - | 3 | 375 | |||
BC118631.1 | - | 2 | 239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
