siRNA Details
Gene Symbol: RPL34-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026968.1 | 4 | 2930 | |||
GenBank mRNA | AK094992.1 | - | 4 | 2873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
