siRNA Details
Gene Symbol: INTS4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027392.2 | 4 | 485 | |||
GenBank mRNA | BC111554.1 | - | 4 | 478 | 40024359 |
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 10 | 1116 | ||
XM_011545352.2 | XP_011543654.1 | 11 | 1285 | |||
XM_011545353.2 | XP_011543655.1 | 10 | 1294 | |||
XM_017018560.1 | XP_016874049.1 | 10 | 1149 | |||
GenBank mRNA | AK290866.1 | - | 10 | 1112 | ||
AL834171.1 | - | 10 | 1059 | |||
BC008013.1 | - | 6 | 569 | |||
BC009995.2 | - | 10 | 1111 | 4129693 | ||
BC015664.2 | - | 9 | 1009 | 4554197 |
Gene Symbol: INTS4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057766.1 | - | 4 | 478 |
Gene Symbol: LOC101929322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516746.2 | XP_011515048.1 | 4 | 779 | ||
XM_011516747.2 | XP_011515049.1 | 4 | 708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
