siRNA Details
Gene Symbol: INTS4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027392.2 | 6 | 886 | |||
GenBank mRNA | BC111554.1 | - | 6 | 879 | 40024359 |
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 12 | 1517 | ||
XM_011545352.2 | XP_011543654.1 | 13 | 1686 | |||
XM_011545353.2 | XP_011543655.1 | 12 | 1695 | |||
XM_017018560.1 | XP_016874049.1 | 12 | 1550 | |||
GenBank mRNA | AK290866.1 | - | 12 | 1513 | ||
AL834171.1 | - | 12 | 1460 | |||
BC008013.1 | - | 8 | 970 | |||
BC009859.2 | - | 2 | 176 | |||
BC009995.2 | - | 12 | 1512 | 4129693 | ||
BC015664.2 | - | 11 | 1410 | 4554197 |
Gene Symbol: INTS4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057766.1 | - | 6 | 879 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
