siRNA Details
Gene Symbol: LOC286359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097847.1 | - | 1 | 411 | ||
BC105125.1 | - | 1 | 323 | |||
BC112056.1 | - | 1 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
