siRNA Details
Gene Symbol: LOC286359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026847.1 | 6 | 620 | |||
GenBank mRNA | AK097847.1 | - | 1 | 1032 | ||
AK310296.1 | - | 6 | 620 | |||
AL833510.1 | - | 4 | 2061 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
