siRNA Details
Gene Symbol: KCTD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198353.2 | NP_938167.1 | 2 | 1438 | ||
XM_011513690.2 | XP_011511992.1 | 3 | 1600 | |||
GenBank mRNA | AK123347.1 | - | 2 | 1438 | ||
AK298831.1 | - | 3 | 907 | |||
BC132743.1 | - | 2 | 1212 | 40146765 | ||
BC136793.1 | - | 2 | 1212 | 9020783 |
