siRNA Details
Gene Symbol: PIKFYVE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178000.1 | NP_001171471.1 | 3, 4 | 482 | ||
NM_015040.3 | NP_055855.2 | 3, 4 | 482 | |||
XM_011510790.1 | XP_011509092.1 | 3, 4 | 482 | |||
XM_017003568.1 | XP_016859057.1 | 3, 4 | 386 | |||
XM_017003569.1 | XP_016859058.1 | 3, 4 | 386 | |||
GenBank mRNA | AK127769.1 | - | 3, 4 | 386 | ||
AY457063.1 | - | 3, 4 | 486 | |||
BC017736.2 | - | 3, 4 | 482 | |||
BC125053.1 | - | 3, 4 | 471 | |||
EU794609.1 | - | 3, 4 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
