siRNA Details
Gene Symbol: MACROD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC101216.1 | - | 4 | 622 | ||
BC101217.1 | - | 4 | 622 | |||
BC101218.1 | - | 4 | 622 | |||
BC101219.1 | - | 4 | 622 | |||
BC126936.1 | - | 4 | 622 | |||
BC128036.1 | - | 4 | 622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
