siRNA Details
Gene Symbol: DYX1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130810.3 | NP_570722.2 | 9 | 1477 | ||
GenBank mRNA | AF337549.1 | - | 9 | 1477 | ||
AK302316.1 | - | 7 | 889 |
Gene Symbol: DYX1C1-CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037923.1 | 8 | 1364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
