siRNA Details
Gene Symbol: DYX1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033559.2 | NP_001028731.1 | 9 | 1586 | ||
NM_130810.3 | NP_570722.2 | 10 | 1692 | |||
GenBank mRNA | AF337549.1 | - | 10 | 1692 | ||
AK095201.1 | - | 8 | 1260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
