siRNA Details
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018050.3 | NP_001018060.1 | 3, 4 | 682 | ||
NM_001282884.1 | NP_001269813.1 | 4, 5 | 410 | |||
NM_001282885.1 | NP_001269814.1 | 4, 5 | 369 | |||
NM_138338.4 | NP_612211.1 | 4, 5 | 376 | |||
GenBank mRNA | AB051452.1 | - | 3, 4 | 537 | ||
AK026524.1 | - | 4, 5 | 356 | |||
AY092087.1 | - | 4, 5 | 354 | |||
BC088367.1 | - | 3, 4 | 545 | 30385483 | ||
BU173504.1 | - | 3, 4 | 324 | |||
CR456459.1 | - | 4, 5 | 410 | |||
DA449644.1 | - | 4, 5 | 391 | |||
DA504810.1 | - | 4, 5 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
