siRNA Details
Gene Symbol: KCTD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138444.3 | NP_612453.1 | 1 | 2959 | ||
GenBank mRNA | AF359381.1 | - | 1 | 2944 | ||
AK057635.1 | - | 1 | 608 | |||
AK128363.1 | - | 2 | 2248 | |||
BC013764.1 | - | 1 | 2790 | 4521096 |
