siRNA Details
Gene Symbol: MS4A6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139249.2 | NP_640342.1 | 2 | 244 | ||
GenBank mRNA | AF333758.1 | - | 2 | 295 | ||
AF354931.1 | - | 2 | 244 | |||
AF354932.1 | - | 2 | 128 | |||
AF354933.1 | - | 2 | 184 | |||
BC034588.1 | - | 2 | 158 | |||
BC069322.1 | - | 2 | 244 | 7262144 | ||
BC101783.1 | - | 2 | 304 | 8069289 |
