siRNA Details
Gene Symbol: C6orf136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109938.2 | NP_001103408.1 | 3 | 702 | ||
NM_001161376.1 | NP_001154848.1 | 3 | 1245 | |||
NM_145029.3 | NP_659466.2 | 2 | 300 | |||
XM_011514384.1 | XP_011512686.1 | 4 | 794 | |||
GenBank mRNA | AK127564.1 | - | 3 | 662 | ||
BC016167.1 | - | 3 | 853 | 3509511 | ||
BC064397.1 | - | 3 | 863 | |||
BM695485.1 | - | 2 | 175 | |||
BP255323.1 | - | 3 | 509 |
