siRNA Details
Gene Symbol: OARD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145063.2 | NP_659500.1 | 6 | 889 | ||
GenBank mRNA | AJ420538.1 | - | 6 | 1025 | ||
AK290939.1 | - | 6 | 892 | |||
BC011709.2 | - | 6 | 889 | 3942960 | ||
BC021579.1 | - | 5 | 839 |
