siRNA Details
Gene Symbol: WDR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145241.4 | NP_660284.1 | 3, 4 | 414 | ||
GenBank mRNA | BC012352.1 | - | 3, 4 | 346 | 4634445 | |
DA305733.1 | - | 3, 4 | 337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
