siRNA Details
Gene Symbol: RDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163120.1 | NP_001156592.1 | 4 | 577 | ||
NM_001163121.1 | NP_001156593.1 | 6 | 832 | |||
NM_001163122.1 | NP_001156594.1 | 5 | 731 | |||
NM_001163124.1 | NP_001156596.1 | 3 | 476 | |||
NM_001163130.1 | NP_001156602.1 | 6 | 830 | |||
NM_145654.3 | NP_663629.1 | 7 | 931 | |||
NR_027996.1 | 5 | 582 | ||||
NR_027997.1 | 6 | 762 | ||||
NR_027998.1 | 5 | 663 | ||||
NR_027999.1 | 4 | 562 | ||||
NR_028000.1 | 5 | 661 | ||||
GenBank mRNA | AB080728.1 | - | 7 | 909 | ||
BC127190.1 | - | 7 | 922 | 40036784 | ||
BX376188.2 | - | 4 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
