siRNA Details
Gene Symbol: ZNF69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321163.1 | NP_001308092.1 | 5 | 1221 | ||
NM_021915.3 | NP_068734.1 | 5 | 1179 | |||
GenBank mRNA | AK123536.1 | - | 2 | 1474 | ||
BC051309.1 | - | 5 | 1108 | 5812091 |
Gene Symbol: ZNF440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152357.2 | NP_689570.2 | 4 | 3717 | ||
XM_005259731.3 | XP_005259788.1 | 4 | 3777 | |||
XM_017026253.1 | XP_016881742.1 | 3 | 4102 | |||
XM_017026254.1 | XP_016881743.1 | 3 | 3595 | |||
GenBank mRNA | AK092559.1 | - | 1 | 1813 | ||
AK095252.1 | - | 4 | 3718 | |||
BC035760.1 | - | 4 | 3699 | 5556302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
