siRNA Details
Gene Symbol: MROH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152503.5 | NP_689716.4 | 17 | 2410 | ||
GenBank mRNA | AL833864.1 | - | 9 | 1154 | ||
BC030006.1 | - | 17 | 2387 | 4822338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
