siRNA Details
Gene Symbol: LINC01600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131168.1 | 3 | 1431 | |||
GenBank mRNA | AK056496.1 | - | 3 | 1431 | ||
BC104007.1 | - | 1 | 1215 | 40024375 | ||
BC104008.1 | - | 1 | 1215 | 40024376 | ||
BC104009.1 | - | 1 | 1215 | 40024379 |
