siRNA Details
Gene Symbol: TRAPPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003274.4 | NP_003265.3 | 23 | 5863 | ||
XM_005261168.4 | XP_005261225.1 | 23 | 5733 | |||
XM_011529714.2 | XP_011528016.1 | 24 | 5967 | |||
XM_011529715.2 | XP_011528017.1 | 24 | 5844 | |||
XM_011529716.2 | XP_011528018.1 | 23 | 5820 | |||
XM_011529719.2 | XP_011528021.1 | 22 | 5523 | |||
XM_011529721.2 | XP_011528023.1 | 21 | 5340 | |||
XM_011529722.2 | XP_011528024.1 | 21 | 5306 | |||
XM_011529723.2 | XP_011528025.1 | 20 | 5203 | |||
XM_011529727.2 | XP_011528029.1 | 20 | 5208 | |||
XM_011529728.2 | XP_011528030.1 | 21 | 5489 | |||
XM_011529729.2 | XP_011528031.1 | 20 | 5113 | |||
XM_017028454.1 | XP_016883943.1 | 22 | 5709 | |||
XM_017028455.1 | XP_016883944.1 | 16 | 4709 | |||
GenBank mRNA | AL833668.1 | - | 6 | 4719 | ||
U61500.1 | - | 23 | 5847 |
Gene Symbol: LOC102724200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546082.2 | XP_011544384.2 | 4 | 2943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
