siRNA Details
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003276.2 | NP_003267.1 | 3, 4 | 868 | ||
XM_017019914.1 | XP_016875403.1 | 3, 4 | 560 | |||
GenBank mRNA | AK315221.1 | - | 3, 4 | 798 | ||
S76736.1 | - | 3, 4 | 773 | |||
U09086.1 | - | 3, 4 | 777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
