siRNA Details
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146261.2 | NP_001139733.1 | 10 | 2138 | ||
NM_001146262.2 | NP_001139734.1 | 9 | 2017 | |||
NM_001146264.2 | NP_001139736.1 | 9 | 2081 | |||
NM_001256006.1 | NP_001242935.1 | 9 | 2165 | |||
NM_153262.3 | NP_694994.2 | 8 | 1960 | |||
NR_027459.2 | 9 | 1894 | ||||
XM_006711262.3 | XP_006711325.1 | 8 | 3196 | |||
XM_017000931.1 | XP_016856420.1 | 7 | 3139 | |||
XM_017000932.1 | XP_016856421.1 | 10 | 2212 | |||
XM_017000933.1 | XP_016856422.1 | 10 | 2075 | |||
XM_017000934.1 | XP_016856423.1 | 7 | 1871 | |||
XM_017000935.1 | XP_016856424.1 | 8 | 1754 | |||
GenBank mRNA | AJ617624.1 | - | 8 | 2046 | ||
AJ617625.1 | - | 9 | 1808 | |||
AJ617626.1 | - | 6 | 1714 | |||
AJ617627.1 | - | 5 | 1452 | |||
AK091517.1 | - | 8 | 1883 | |||
BC130323.1 | - | 10 | 2071 | 40146354 | ||
BC144154.1 | - | 9 | 2010 | |||
BC144155.1 | - | 9 | 1950 | |||
BC144157.1 | - | 10 | 2071 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
