siRNA Details
Gene Symbol: ADGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153840.3 | NP_722582.2 | 10, 11 | 1340 | ||
XM_011514469.1 | XP_011512771.1 | 7, 8 | 996 | |||
XM_017010735.1 | XP_016866224.1 | 7, 8 | 1098 | |||
XM_017010736.1 | XP_016866225.1 | 8, 9 | 1073 | |||
GenBank mRNA | AB045123.1 | - | 10, 11 | 1294 | ||
AK055208.1 | - | 300 | ||||
AY140952.1 | - | 4, 5 | 533 | |||
BC029358.1 | - | 9, 10 | 1165 | |||
CR627234.1 | - | 8, 9 | 7389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
