siRNA Details
Gene Symbol: PGBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170725.2 | NP_733843.1 | 3 | 763 | ||
XM_005270333.1 | XP_005270390.1 | 4 | 862 | |||
XM_011544159.2 | XP_011542461.1 | 3 | 764 | |||
XM_011544161.2 | XP_011542463.1 | 3 | 1704 | |||
GenBank mRNA | AK298248.1 | - | 4 | 863 | ||
BC063785.1 | - | 3 | 768 | 6173868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
