siRNA Details
Gene Symbol: SLC25A26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164796.1 | NP_001158268.1 | 9 | 1319 | ||
NM_173471.3 | NP_775742.4 | 11 | 1851 | |||
NR_028475.1 | 10 | 1404 | ||||
XM_011533327.2 | XP_011531629.1 | 10 | 1214 | |||
XM_017005674.1 | XP_016861163.1 | 8 | 1026 | |||
GenBank mRNA | BC012852.1 | - | 9 | 1038 | 3860705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
