siRNA Details
Gene Symbol: C10orf126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278522.1 | NP_001265451.1 | 3, 4 | 348 | ||
GenBank mRNA | BC033839.1 | - | 3, 4 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
